Skip to Content
MilliporeSigma
All Photos(1)

Documents

37277

Sigma-Aldrich

(+)-Dihydrocarveol

purum, mixture of isomers, ≥95.0% (sum of enantiomers, GC)

Synonym(s):

(1S,2S,5S)-5-Isopropenyl-2-methylcyclohexanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
Beilstein:
5729399
EC Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.22

grade

purum

Assay

≥95.0% (sum of enantiomers, GC)

optical activity

[α]20/D +20±1°, neat

composition

n-, ~75%
iso-, ~6%
neo-, ~3%
neoiso-, ~13%

refractive index

n20/D 1.476-1.479

density

0.926 g/mL at 20 °C (lit.)

SMILES string

C[C@H]1CC[C@@H](C[C@@H]1O)C(C)=C

InChI

1S/C10H18O/c1-7(2)9-5-4-8(3)10(11)6-9/h8-11H,1,4-6H2,2-3H3/t8-,9-,10-/m0/s1

InChI key

KRCZYMFUWVJCLI-GUBZILKMSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

194.0 °F - closed cup

Flash Point(C)

90 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service