Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

344613

Sigma-Aldrich

Cobalt(II) acetylacetonate hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Co(C5H7O2)2 · xH2O
CAS Number:
Molecular Weight:
257.15 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

form

powder or crystals

composition

Degree of hydration, ~2

reaction suitability

core: cobalt

SMILES string

O.CC(=O)\C=C(\C)O[Co]O\C(C)=C/C(C)=O

InChI

1S/2C5H8O2.Co.H2O/c2*1-4(6)3-5(2)7;;/h2*3,6H,1-2H3;;1H2/q;;+2;/p-2/b2*4-3-;;

InChI key

CCSRIPNIKKQYHL-SUKNRPLKSA-L

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Dam. 1 - Repr. 1A - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service