Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

341061

Sigma-Aldrich

3-Chlorothiophene

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H3ClS
CAS Number:
Molecular Weight:
118.58
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

form

liquid

impurities

<2% N,N-dimethylformamide

refractive index

n20/D 1.553 (lit.)

bp

137-139 °C (lit.)

density

1.276 g/mL at 25 °C (lit.)

SMILES string

Clc1ccsc1

InChI

1S/C4H3ClS/c5-4-1-2-6-3-4/h1-3H

InChI key

QUBJDMPBDURTJT-UHFFFAOYSA-N

General description

Electrochemical homopolymerization of 3-chlorothiophene and copolymerization with 3-methylthiophene in ionic liquid has been investigated by potentiodynamic and galvanostat methods.

Application

3-Chlorothiophene was used in the synthesis of poly(3-chlorothiophene) films by direct oxidation in mixed electrolytes of boron trifluoride diethyl etherate and trifluroacetic acid.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Repr. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

98.6 °F - closed cup

Flash Point(C)

37 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Low potential electrochemical polymerization of 3-chlorothiophene in mixed electrolytes of boron trifluoride diethyl etherate and trifluoroacetic acid.
Xu J, et al.
Journal of Electroanalytical Chemistry, 514(1), 16-25 (2001)
Victor Abrahamsson et al.
Journal of chromatography. A, 1501, 89-98 (2017-04-30)
Fouling tendencies of a series of gas condensates were evaluated using comprehensive two-dimensional gas chromatography with flame ionization detection and sulfur chemiluminescence detection. A pixel-based approach was applied in order to identify parts of the chromatograms which were associated with
Electrochemical synthesis, characterization, and electrochromic properties of poly (3-chlorothiophene) and its copolymer with 3-methylthiophene in a room temperature ionic liquid.
Pang Y, et al.
Electrochemical Communications, 8(!1), 1757-1763 (2006)
Hilal E Toraman et al.
Journal of chromatography. A, 1359, 237-246 (2014-07-30)
The detailed compositional characterization of plastic waste pyrolysis oil was performed with comprehensive two-dimensional GC (GC×GC) coupled to four different detectors: a flame ionization detector (FID), a sulfur chemiluminescence detector (SCD), a nitrogen chemiluminescence detector (NCD) and a time of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service