Skip to Content
MilliporeSigma
All Photos(1)

Documents

324485

Sigma-Aldrich

Dichloromethyldiisopropoxyborane

97%

Synonym(s):

(Dichloromethyl)diisopropoxyborane, Diisopropyl (dichloromethyl)boronate, Diisopropyl dichloromethaneboronate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2CHB[OCH(CH3)2]2
CAS Number:
Molecular Weight:
212.91
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

reaction suitability

reagent type: reductant

density

1.019 g/mL at 25 °C (lit.)

SMILES string

CC(C)OB(OC(C)C)C(Cl)Cl

InChI

1S/C7H15BCl2O2/c1-5(2)11-8(7(9)10)12-6(3)4/h5-7H,1-4H3

InChI key

XHOSKSMGPJTTSE-UHFFFAOYSA-N

Application

Reactant for:
  • Formation of carbon-carbon and carbon-heteroatom bonds
  • Solid phase synthesis of aminoboronic acids as potent inhibitors of the hepatitis C virus NS3 proteinase

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

104.0 °F - closed cup

Flash Point(C)

40 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organometallics, 5, 1465-1465 (1986)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service