Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

305324

Sigma-Aldrich

(Formylmethyl)triphenylphosphonium chloride

97%

Synonym(s):

2-Oxoethyltriphenylphosphonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCOCH2P(Cl)(C6H5)3
CAS Number:
Molecular Weight:
340.78
Beilstein:
4060813
EC Number:
MDL number:
UNSPSC Code:
12352000

Assay

97%

mp

209-212 °C (dec.) (lit.)

SMILES string

[Cl-].O=CC[P+](c1ccccc1)(c2ccccc2)c3ccccc3

InChI

1S/C20H18OP.ClH/c21-16-17-22(18-10-4-1-5-11-18,19-12-6-2-7-13-19)20-14-8-3-9-15-20;/h1-16H,17H2;1H/q+1;/p-1

InChI key

RVEJRPJGKXTQIF-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for:
  • Hydroazidation reactions in preparation of a-azido alcohols
  • Preparation of prodrug (2-hydroxyamino-vinyl)-triphenyl-phosphonium (HVTP) inhibitor of peroxidase activity and apoptosis
  • Intramolecular [4 + 3] cycloaddition reactions

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service