Skip to Content
MilliporeSigma
All Photos(1)

Documents

293806

Sigma-Aldrich

2-Ethyl-3-methoxypyrazine

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H10N2O
CAS Number:
Molecular Weight:
138.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

refractive index

n20/D 1.502 (lit.)

bp

90 °C/40 mmHg (lit.)

density

1.049 g/mL at 25 °C (lit.)

SMILES string

CCc1nccnc1OC

InChI

1S/C7H10N2O/c1-3-6-7(10-2)9-5-4-8-6/h4-5H,3H2,1-2H3

InChI key

DPCILIMHENXHQX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Ethyl-3-methoxypyrazine has been employed as internal standard for determination of taste and odour causing compounds using the granular adsorbent, Ambersorb 572, by gas chromatography-high resolution mass spectrometry.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

156.2 °F - closed cup

Flash Point(C)

69 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J P Palmentier et al.
The Analyst, 126(6), 840-845 (2001-07-12)
A method for the analysis of six taste and odour causing compounds in aqueous samples using the granular adsorbent, Ambersorb 572, and gas chromatography-high resolution mass spectrometry (GC-HRMS) has been developed. This method for the determination of geosmin, 2-methylisoborneol (2-MIB)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service