Skip to Content
MilliporeSigma
All Photos(1)

Documents

28894

Sigma-Aldrich

cis,cis-1,9-Cyclohexadecadiene

≥97.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H28
CAS Number:
Molecular Weight:
220.39
Beilstein:
1363605
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0%

bp

115 °C/0.001 mmHg (lit.)

SMILES string

C1CCCC=CCCCCCCC=CCC1

InChI

1S/C16H28/c1-2-4-6-8-10-12-14-16-15-13-11-9-7-5-3-1/h1-2,15-16H,3-14H2/b2-1-,16-15-

InChI key

QCSDLMPAQMQZOE-QSWGICILSA-N

Application

1,9-Cyclohexadecadiene was used in the synthesis of muscone and exaltone.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of racemic muscone and cyclopentadecanone (exaltone) from 1, 9-cyclohexadecadiene.
Mookherjee BD, et al.
The Journal of Organic Chemistry, 36(22), 3266-3270 (1971)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service