Skip to Content
MilliporeSigma
All Photos(1)

Documents

28706

Sigma-Aldrich

trans-Cyclodecene

≥98.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18
CAS Number:
Molecular Weight:
138.25
Beilstein:
2036982
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥98.0% (GC)

refractive index

n20/D 1.484

density

0.867 g/mL at 20 °C (lit.)

SMILES string

C1CCCC\C=C\CCC1

InChI

1S/C10H18/c1-2-4-6-8-10-9-7-5-3-1/h1-2H,3-10H2/b2-1+

InChI key

UCIYGNATMHQYCT-OWOJBTEDSA-N

Application

trans-Cyclodecene was used as standard of reference for the quantitative infrared analysis of mixtures of cis- and trans-cyclodecene.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

143.6 °F - closed cup

Flash Point(C)

62 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Many-membered Carbon Rings. IX. trans-Elimination in Thermal Decomposition of Cyclodecyl Esters1.
Blomquist AT and Goldstein A.
Journal of the American Chemical Society, 77(4), 1001-1003 (1955)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service