Skip to Content
MilliporeSigma
All Photos(1)

Documents

276162

Sigma-Aldrich

1,3-Dibromo-1-propene, mixture of cis and trans

98%

Synonym(s):

1,3-Dibromo-2-propene, 1,3-Dibromopropene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrCH2CH=CHBr
CAS Number:
Molecular Weight:
199.87
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.5573 (lit.)

bp

155-156 °C (lit.)

density

2 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

BrC\C=C\Br

InChI

1S/C3H4Br2/c4-2-1-3-5/h1-2H,3H2/b2-1+

InChI key

JWQMKMSVOYQICF-OWOJBTEDSA-N

Application

1,3-Dibromo-1-propene is a synthetic building block and has been used in the preparation of 3-bromo-3-phenoxy-1-palladacyclobutane.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

80.6 °F - closed cup

Flash Point(C)

27 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mechanism of Nucleophilic Attack on 1-and 2-Bromo (p-allyl) palladium Complexes1.
Organ MG, et al.
Journal of the American Chemical Society, 120(36), 9283-9290 (1998)
Palladium-catalyzed direct functionalization of imidazolinone: synthesis of dibromophakellstatin.
Jianming Lu et al.
Journal of the American Chemical Society, 129(25), 7768-7769 (2007-06-02)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service