Skip to Content
MilliporeSigma
All Photos(2)

Documents

255971

Sigma-Aldrich

Iron(II) oxalate dihydrate

≥99.99% trace metals basis

Synonym(s):

Ferrous oxalate dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FeC2O4 · 2H2O
CAS Number:
Molecular Weight:
179.89
Beilstein:
3757620
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

≥99.99% trace metals basis

form

powder

impurities

≤100.0 ppm Trace Metal Analysis

density

2.28 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

[H]O[H].[H]O[H].O=C1O[Fe]OC1=O

InChI

1S/C2H2O4.Fe.2H2O/c3-1(4)2(5)6;;;/h(H,3,4)(H,5,6);;2*1H2/q;+2;;/p-2

InChI key

NPLZZSLZTJVZSX-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service