Skip to Content
MilliporeSigma
All Photos(2)

Documents

250120

Sigma-Aldrich

Ethyl 3-ethoxyacrylate

98%

Synonym(s):

Ethyl 3-ethoxy-2-propenoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2OCH=CHCOOCH2CH3
CAS Number:
Molecular Weight:
144.17
Beilstein:
1721612
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.446 (lit.)

bp

195-196 °C (lit.)

density

0.998 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCO\C=C\C(=O)OCC

InChI

1S/C7H12O3/c1-3-9-6-5-7(8)10-4-2/h5-6H,3-4H2,1-2H3/b6-5+

InChI key

ITQFPVUDTFABDH-AATRIKPKSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

168.8 °F - closed cup

Flash Point(C)

76 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

New preparations of ethyl 3, 3-diethoxypropionate and ethoxycarbonylmalondialdehyde. Copper (I) catalyzed acetal formation from a conjugated triple bond.
Bertz SH, et al.
The Journal of Organic Chemistry, 47(11), 2216-2217 (1982)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service