Skip to Content
MilliporeSigma
All Photos(1)

Documents

238635

Sigma-Aldrich

IGEPAL® CO-210

average Mn ~308

Synonym(s):

Polyoxyethylene (2) nonylphenyl ether, branched

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H4O)n · C15H24O, n~1.5
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

mol wt

average Mn ~308

refractive index

n20/D 1.508 (lit.)

density

0.96 g/mL at 25 °C (lit.)

HLB

4

SMILES string

CCCCCCCCCc1ccc(OCCOCCO)cc1

Gene Information

rat ... Ar(24208)

Legal Information

IGEPAL is a registered trademark of Solvay

comparable product

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Markus A Lill et al.
Journal of medicinal chemistry, 48(18), 5666-5674 (2005-09-02)
We investigated the influence of induced fit of the androgen receptor binding pocket on free energies of ligand binding. On the basis of a novel alignment procedure using flexible docking, molecular dynamics simulations, and linear-interaction energy analysis, we simulated the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service