Skip to Content
MilliporeSigma
All Photos(1)

Documents

229385

Sigma-Aldrich

Violamine R

Dye content 60 %

Synonym(s):

Acid Violet 9

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H25N2NaO6S
CAS Number:
Molecular Weight:
612.63
Colour Index Number:
45190
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

form

powder

composition

Dye content, 60%

solubility

water: 1%

λmax

529 nm

ε (extinction coefficient)

≥27000 at 525-531 nm in water at 0.01 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].Cc1ccccc1Nc2ccc3c(OC4=CC(\C=CC4=C3c5ccccc5C(O)=O)=N\c6ccc(cc6C)S([O-])(=O)=O)c2

InChI

1S/C34H26N2O6S.Na/c1-20-7-3-6-10-29(20)35-22-11-14-27-31(18-22)42-32-19-23(36-30-16-13-24(17-21(30)2)43(39,40)41)12-15-28(32)33(27)25-8-4-5-9-26(25)34(37)38;/h3-19,35H,1-2H3,(H,37,38)(H,39,40,41);/q;+1/p-1/b36-23+;

InChI key

FRCXWIMQWXXKSC-CWWQQMKRSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Parisi et al.
Bollettino della Societa italiana di biologia sperimentale, 58(11), 690-694 (1982-06-15)
From the examination of the visible and ultraviolet spectra, together with the excitation and fluorescence spectra of the chromatographically pure samples of rhodamine B, rhodamine 6G and violamine R, it is possible to establish the nature of the chromophores in
G Parisi et al.
Bollettino della Societa italiana di biologia sperimentale, 58(11), 683-689 (1982-06-15)
Rhodamines, commonly used in microscopic studies, are basic dyes derived from xanthene. They have recently been used for the quantitative analysis of important cations, as well as in the vital staining of mitochondria of normal or pathological cells and also

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service