Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

221430

Sigma-Aldrich

1,3,5-Triethylhexahydro-1,3,5-triazine

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H21N3
CAS Number:
Molecular Weight:
171.28
EC Number:
MDL number:
UNSPSC Code:
12352100

Assay

95%

impurities

<5% water

refractive index

n20/D 1.4595 (lit.)

bp

207-208 °C (lit.)

density

0.894 g/mL at 25 °C (lit.)

SMILES string

CCN1CN(CC)CN(CC)C1

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

158.0 °F - closed cup

Flash Point(C)

70 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Linda E Winfield et al.
Ecotoxicology (London, England), 13(4), 335-347 (2004-09-04)
Soils contaminated with explosive materials like hexahydro-1,3,5-trinitro-1,3,5-triazine (RDX) is a concern nation-wide on military installations and sites where explosives are manufactured, stored, or disposed. Terrestrial plants are a vital group of receptor organism, yet limited published information is available on
Ronald E Baynes et al.
Toxicology letters, 159(3), 252-260 (2005-06-28)
Triazine is often added to cutting-fluid formulations in the metal-machining industry as a preservative. Trichloroethylene (TCE) is a solvent used for cleaning the cutting fluid or oil from the metal product. The purpose of this study was to examine the
Ping Gong et al.
Environmental toxicology and chemistry, 26(5), 954-959 (2007-05-25)
Hexanitrohexaazaisowurtzitane (CL-20) and hexahydro-1,3,5-trinitro-1,3,5-triazine (RDX), both energetic compounds, share some degree of structural similarity. A noninvasive electrophysiological technique was employed to assess the impacts of acute sublethal exposures on impulse conduction in medial (MGF) and lateral (LGF) giant nerve fiber
P Gong et al.
Environmental toxicology and chemistry, 20(5), 947-951 (2001-05-08)
Although hexahydro-1,3,5-trinitro-1,3,5-triazine (also called RDX or hexogen) is a potentially toxic explosive compound that persists in soil, its ecotoxicological effects on soil organisms have rarely been assessed. In this study, two uncontaminated garden soils were spiked with 10 to 12,500

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service