Skip to Content
MilliporeSigma
All Photos(1)

Documents

216534

Supelco

Amberlite IRC120 H, hydrogen form

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
23151817

vapor density

>1 (vs air)

vapor pressure

17 mmHg ( 20 °C)

description

cross-linkage 8%
cross-linkage 8%
hydrogen form
moisture 53-58%

form

beads

autoignition temp.

800 °F

parameter

121 °C max. temp.

technique(s)

LPLC: suitable

matrix

styrene-divinylbenzene (gel)

matrix active group

sulfonic acid

particle size

620-830 μm

operating pH

0-14

capacity

1.8 meq/mL by wetted bed volume

separation technique

cation exchange

InChI

1S/C10H10.C8H8O3S/c1-3-9-7-5-6-8-10(9)4-2;1-2-7-5-3-4-6-8(7)12(9,10)11/h3-8H,1-2H2;2-6H,1H2,(H,9,10,11)

InChI key

SIWVGXQOXWGJCI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Strongly acidic cation exchange resin suitable for a wide variety of chemical process applications, removal of amino acids at low pH, USP potassium methods, etc..

Features and Benefits

Strongly acidic gel-type resin with sulfonic acid functionality which is useful in catalytic applications.

Legal Information

Amberlite is a trademark of DuPont de Nemours, Inc.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service