Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

192902

Sigma-Aldrich

Z-N-Me-Aib-OH

98%

Synonym(s):

Z-N,2-Dimethylalanine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2OCON(CH3)C(CH3)2CO2H
CAS Number:
Molecular Weight:
251.28
Beilstein:
3347632
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

reaction suitability

reaction type: solution phase peptide synthesis

mp

135-138 °C (lit.)

application(s)

peptide synthesis

SMILES string

CN(C(=O)OCc1ccccc1)C(C)(C)C(O)=O

InChI

1S/C13H17NO4/c1-13(2,11(15)16)14(3)12(17)18-9-10-7-5-4-6-8-10/h4-8H,9H2,1-3H3,(H,15,16)

InChI key

QCDSXBNEBTVGTP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service