Skip to Content
MilliporeSigma
All Photos(1)

Documents

183628

Sigma-Aldrich

3,4-Dichlorobenzyl alcohol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3CH2OH
CAS Number:
Molecular Weight:
177.03
Beilstein:
2082362
EC Number:
MDL number:
UNSPSC Code:
12352103

Assay

98%

bp

148-151 °C (lit.)

mp

35-38 °C (lit.)

SMILES string

OCc1ccc(Cl)c(Cl)c1

InChI

1S/C7H6Cl2O/c8-6-2-1-5(4-10)3-7(6)9/h1-3,10H,4H2

InChI key

FVJIUQSKXOYFKG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

>235.4 °F - closed cup

Flash Point(C)

> 113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service