Skip to Content
MilliporeSigma
All Photos(1)

Documents

160342

Sigma-Aldrich

2-Fluoroethanol

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FCH2CH2OH
CAS Number:
Molecular Weight:
64.06
Beilstein:
1730857
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

16 mmHg ( 20 °C)

Assay

95%

form

liquid

refractive index

n20/D 1.365 (lit.)

bp

103 °C/757 mmHg (lit.)

mp

−26.5 °C (lit.)

density

1.091 g/mL at 25 °C (lit.)

SMILES string

OCCF

InChI

1S/C2H5FO/c3-1-2-4/h4H,1-2H2

InChI key

GGDYAKVUZMZKRV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The microwave spectra of two isotopic species of 2-fluoroethanol have been investigated. The Gg′ conformer of 2-fluoroethanol was isolated in low-temperature noble gas matrices. Single-photon IR photolysis of 2-fluoroethanol in solid argon has been investigated.

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Oral - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

93.2 °F - closed cup

Flash Point(C)

34 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kiel D Neumann et al.
Journal of labelled compounds & radiopharmaceuticals, 60(7), 337-342 (2017-04-14)
O-(2-Fluoroethyl)-O-(p-nitrophenyl) methylphosphonate 1 is an organophosphate cholinesterase inhibitor that creates a phosphonyl-serine covalent adduct at the enzyme active site blocking cholinesterase activity in vivo. The corresponding radiolabeled O-(2-[
Microwave Spectrum and Intramolecular Hydrogen Bonding in 2-Fluoroethanol.
Buckton KS and Azrak RG.
J. Chem. Phys. , 52(11), 5652-5655 (1970)
Site-selective infrared photoisomerizations of 2-fluoroethanol in low-temperature matrices
Pourcin J, et al.
Journal of Molecular Spectroscopy, 109(1) (1985)
Infrared laser photoisomerization of 2-fluoroethanol in argon.
Hoffman III WF and Shirk JS.
Chemical Physics, 78(3), 331-338 (1983)
Nicole Borho et al.
Journal of the American Chemical Society, 130(18), 5916-5921 (2008-04-12)
The conformational isomerism of the propylene oxide (PO)...2-fluoroethanol (FE) complex has been investigated using molecular beam Fourier-transform microwave spectroscopy complemented with high level ab initio calculations. Rotational transitions of three different binary conformers have been observed experimentally. On the basis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service