Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

154229

Sigma-Aldrich

4-Methoxy-1-naphthonitrile

99%

Synonym(s):

1-Cyano-4-methoxynaphthalene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC10H6CN
CAS Number:
Molecular Weight:
183.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

99%

form

liquid

mp

100-102 °C (lit.)

SMILES string

COc1ccc(C#N)c2ccccc12

InChI

1S/C12H9NO/c1-14-12-7-6-9(8-13)10-4-2-3-5-11(10)12/h2-7H,1H3

InChI key

FRCZUGOIGNQOID-UHFFFAOYSA-N

Application

4-Methoxy-1-naphthonitrile was used in the preparation of 1(endo),8b-dicyano-4-methoxy-1,2,2a-trihydrocyclobuta[a]naphthalene. It was also used to synthesize substituted azocin-2(1H)-ones.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The synthesis of azocine derivatives by the photocycloaddition of phenols to 1-naphthonitrile and 4-methoxy-1-naphthonitrile.
Al-Jalal NA.
Journal of Photochemistry and Photobiology A: Chemistry, 54(1), 99-104 (1990)
The photocycloaddition of 4-methoxy-1-naphthonitrile and 2-methoxy-1-naphthonitrile with acrylonitrile and 3, 4-dihydro-2H-pyran: Facile additions in sunlight.
Journal of Photochemistry and Photobiology A: Chemistry, 51(3), 429-436 (1990)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service