Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

15125

Sigma-Aldrich

Bis(4-nitrophenyl) phosphate hydrate

≥97.0% (dried material, T)

Synonym(s):

4-Nitrophenyl phosphate monobasic

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(O2NC6H4O)2P(O)OH
CAS Number:
Molecular Weight:
340.18 (anhydrous basis)
Beilstein:
2064857
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0% (dried material, T)

form

powder

storage temp.

2-8°C

SMILES string

O.OP(=O)(Oc1ccc(cc1)[N+]([O-])=O)Oc2ccc(cc2)[N+]([O-])=O

InChI

1S/C12H9N2O8P.H2O/c15-13(16)9-1-5-11(6-2-9)21-23(19,20)22-12-7-3-10(4-8-12)14(17)18;/h1-8H,(H,19,20);1H2

InChI key

AJCYEUQRBLMJDI-UHFFFAOYSA-N

General description

The hydrolysis of Bis(4-nitrophenyl) phosphate hydrate was studied.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hydrolysis of bis (4-nitrophenyl) phosphate catalyzed by metallomicelle made up of the crowned Schiff base complex as synthetic hydrolase.
Jia-qing X, et al.
Journal of Dispersion Science and Technology, 26(6), 693-699 (2005)
Jie Gao et al.
Experimental cell research, 387(2), 111780-111780 (2019-12-25)
Heat shock transcription factor 1 (HSF1) is a highly versatile transcription factor that, in addition to protecting cells against proteotoxic stress, is also critical during diverse developmental processes. Although the functions of HSF1 have received considerable attention, its potential role

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service