Skip to Content
MilliporeSigma
All Photos(1)

Documents

150762

Sigma-Aldrich

Benzamide

zone-refined, purified by sublimation, 99.9%

Synonym(s):

Benzoic acid amide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CONH2
CAS Number:
Molecular Weight:
121.14
Beilstein:
385876
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

zone-refined

Assay

99.9%

purified by

sublimation

mp

125-128 °C (lit.)

SMILES string

NC(=O)c1ccccc1

InChI

1S/C7H7NO/c8-7(9)6-4-2-1-3-5-6/h1-5H,(H2,8,9)

InChI key

KXDAEFPNCMNJSK-UHFFFAOYSA-N

Gene Information

human ... PARP1(142)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Inhibits poly(ADP-ribose) polymerase (PARP).

comparable product

Product No.
Description
Pricing

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Muta. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

356.0 °F - closed cup

Flash Point(C)

180 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A P de Jong et al.
Journal of chromatography, 419, 233-242 (1987-08-07)
A universal rapid, sensitive and selective high-performance liquid chromatographic method with UV detection at 230 nm has been developed for the determination of benzamide drugs in human plasma and urine. Sample pretreatment is carried out using solid-phase extraction columns, resulting
Ryoko Chiba et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 19(5), 785-789 (2003-05-29)
A column-switching high-performance liquid chromatographic method with fluorescence detection was developed for the simultaneous determination of four benzamide-type anti-psychotic drugs: sulpiride, tiapride, sultopride and metoclopramide in human serum. In this method, a TSKgel Super-ODS column was used as an analytical
J D Coyle et al.
Journal of pharmaceutical sciences, 76(5), 402-405 (1987-05-01)
We report a reversed-phase high-performance liquid chromatography method for the determination of procainamide (PA) and three of its metabolites, n-acetylprocainamide (NAPA), deethylprocainamide (DEPA), and deethyl-n-acetylprocainamide (DENAPA), in serum and urine. (p-Amino)-n-(2-dipropylaminoethyl)-benzamide was the internal standard. A phenyl column (1.0-mL/min flow
J M Chemnitius et al.
Pharmacological research, 34(1-2), 65-72 (1996-07-01)
Inhibitory effects of the dopamine D2-receptor antagonistic benzamide compound metoclopramide (MCP) on acetylcholinesterase (AChE; EC 3.1.1.7) isoenzymes of both erythrocytes and human caudate nucleus and on human serum cholinesterase (ChE; EC 3.1.1.8) were studied in vitro using a spectrophotometric assay
M H Gschwend et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 831(1-2), 132-139 (2006-01-03)
The development of a validated liquid chromatography-tandem mass spectrometry (LC-MS/MS) method with positive electrospray ionisation (ESI(+)) and multiple reaction monitoring (MRM) for the selective and sensitive bioanalytical determination of amisulpride, a substituted benzamide derivative, in human plasma is described. Plasma

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service