Skip to Content
MilliporeSigma
All Photos(3)

Documents

149349

Sigma-Aldrich

4-(Dimethylamino)benzophenone

98%

Synonym(s):

4-Benzoyl-N ,N -dimethylaniline, p -Benzoyl-N ,N -dimethylaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NC6H4COC6H5
CAS Number:
Molecular Weight:
225.29
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

form

powder

mp

88-90 °C (lit.)

SMILES string

CN(C)c1ccc(cc1)C(=O)c2ccccc2

InChI

1S/C15H15NO/c1-16(2)14-10-8-13(9-11-14)15(17)12-6-4-3-5-7-12/h3-11H,1-2H3

InChI key

BEUGBYXJXMVRFO-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Yu Cai et al.
ACS nano, 11(1), 1054-1063 (2016-12-30)
Herein, a donor-acceptor-donor (D-A-D) structured small molecule (DPP-TPA) is designed and synthesized for photoacoustic imaging (PAI) guided photodynamic/photothermal synergistic therapy. In the diketopyrrolopyrrole (DPP) molecule, a thiophene group is contained to increase the intersystem crossing (ISC) ability through the heavy
Juan Li et al.
Environmental science & technology, 53(18), 10969-10977 (2019-08-15)
Photoinitiators (PIs) are widely used in industrial polymerization and have been detected as emerging contaminants in environmental matrixes. It has been reported that humans are exposed to PIs, but the maternal-fetal transmission of PIs has not been documented. In this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service