Skip to Content
MilliporeSigma
All Photos(1)

Documents

142166

Sigma-Aldrich

Triphenylantimony(V) diacetate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CO2)2Sb(C6H5)3
CAS Number:
Molecular Weight:
471.16
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

97%

reaction suitability

core: antimony
reagent type: catalyst

mp

217-221 °C (lit.)

SMILES string

CC(=O)O[Sb](OC(C)=O)(c1ccccc1)(c2ccccc2)c3ccccc3

InChI

1S/3C6H5.2C2H4O2.Sb/c3*1-2-4-6-5-3-1;2*1-2(3)4;/h3*1-5H;2*1H3,(H,3,4);/q;;;;;+2/p-2

InChI key

MPOFCRCFZPKTCF-UHFFFAOYSA-L

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mio Matsumura et al.
Chemical & pharmaceutical bulletin, 63(2), 130-133 (2015-03-10)
The reaction of triarylantimony diacetates [Ar3Sb(OAc)2] with dialkyl H-phosphites [H-PO(OR)2] in the presence of a Pd(PPh3)4 (5 mol%) catalyst led to the formation of arylphosphonates in moderate to excellent yield under base-free conditions. This reaction is the first example of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service