Skip to Content
MilliporeSigma
All Photos(1)

Documents

141224

Sigma-Aldrich

2-Chloro-6-fluorotoluene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(Cl)F
CAS Number:
Molecular Weight:
144.57
Beilstein:
1932657
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.504 (lit.)

bp

154-156 °C (lit.)

density

1.191 g/mL at 25 °C (lit.)

SMILES string

Cc1c(F)cccc1Cl

InChI

1S/C7H6ClF/c1-5-6(8)3-2-4-7(5)9/h2-4H,1H3

InChI key

FNPVYRJTBXHIPB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Chloro-6-fluorotoluene reaction with SO− -4 has been studied by pulse radiolysis.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

114.8 °F

Flash Point(C)

46 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pulse radiolysis study of the reactions of SO− -4 with some substituted benzenes in aqueous solution.
Merga G, et al.
J. Chem. Soc., Faraday, 90(4), 597-604 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service