Skip to Content
MilliporeSigma
All Photos(1)

Documents

139246

Sigma-Aldrich

1,1,3,3-Tetramethyldisilazane

97%

Synonym(s):

TMDS

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2SiH]2NH
CAS Number:
Molecular Weight:
133.34
Beilstein:
741869
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.404 (lit.)

mp

99-100 °C

density

0.752 g/mL at 25 °C (lit.)

SMILES string

[H][Si](C)(C)N[Si]([H])(C)C

InChI

1S/C4H15NSi2/c1-6(2)5-7(3)4/h5-7H,1-4H3

InChI key

NQCZAYQXPJEPDS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

51.8 °F

Flash Point(C)

11 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tae Hyeon Kim et al.
PloS one, 15(10), e0240219-e0240219 (2020-10-10)
Endoscopic transpapillary gallbladder drainage (ETGBD) has been proposed as an alternative to surgery or percutaneous cholecystostomy in patients with acute calculus cholecystitis (ACC). We aimed to evaluate the safety and efficacy of ETGBD via endoscopic transpapillary gallbladder stenting (ETGBS) or
Saeede Amirighadi et al.
Materials science & engineering. C, Materials for biological applications, 75, 784-790 (2017-04-19)
A simple low-cost method is proposed to fabricate a functionalized multi-wall carbon nanotube (MWCNT) with 1,1,3,3-tetramethyldisilazane (TMDS) molecule. The techniques of scanning electron microscope (SEM) with electron diffraction and energy dispersive X-ray (EDAX) analysis were applied to characterize the functionalized
Kate Butcher et al.
BMC cancer, 18(1), 753-753 (2018-07-23)
Disulfiram (DS), an antialcoholism medicine, demonstrated strong anticancer activity in the laboratory but did not show promising results in clinical trials. The anticancer activity of DS is copper dependent. The reaction of DS and copper generates reactive oxygen species (ROS).
Koji Kosaka et al.
Environmental science & technology, 48(19), 11243-11250 (2014-09-04)
N-Nitrosodimethylamine (NDMA), a potential human carcinogen, is known to be a disinfection byproduct of chloramination and ozonation. NDMA is formed during ozonation at water purification plants in the Yodo River basin, a major drinking water source in western Japan. An
André M Bienfait et al.
Dalton transactions (Cambridge, England : 2003), 43(46), 17324-17332 (2014-10-17)
Trivalent Eu[N(SiHMe2)2]3(THF)2 can easily be synthesized by applying a routine salt metathesis protocol (EuCl3(THF)2 and 3 equiv. of Li[N(SiHMe2)2] in n-hexane) which crystallizes isotypically to its analogues of the rare-earth metal series (space group P21/c). Transsilylamination of Eu[N(SiMe3)2]2(THF)2 with a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service