Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

137111

Sigma-Aldrich

Ethoxyacetic acid

98%

Synonym(s):

O-Ethylglycolic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OCH2COOH
CAS Number:
Molecular Weight:
104.10
Beilstein:
1743037
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.419 (lit.)

bp

97-100 °C/11 mmHg (lit.)

density

1.102 g/mL at 25 °C (lit.)

SMILES string

CCOCC(O)=O

InChI

1S/C4H8O3/c1-2-7-3-4(5)6/h2-3H2,1H3,(H,5,6)

InChI key

YZGQDNOIGFBYKF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethoxyacetic acid is biomarker of exposure to widely used industrial chemicals such as alkyl-substituted ethylene glycols and other aliphatic ethers.

Application

Ethoxyacetic acid is metabolite of glycol ether alkoxy acid and was used to evaluate its toxicity in embryonic stem cell test in mouse. It was used in zebrafish embryotoxicity test to study chemical toxicity after exposure of the complete vertebrate embryo during embryogenesis in ovo.

Biochem/physiol Actions

Ethoxyacetic acid induces testicular toxicity in rats in vivo .

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

206.6 °F - closed cup

Flash Point(C)

97 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Groeseneken et al.
International archives of occupational and environmental health, 61(4), 249-254 (1989-01-01)
A sensitive and specific method for the determination in urine of alkoxyacetic acids, the metabolites of ethylene glycol monoalkyl ethers, was developed by combining the advantages of two previously described methods. The acids were determined gas chromatographically as their pentafluorobenzylesters.
J C Hoflack et al.
Mutation research, 341(4), 281-287 (1995-02-01)
Ethylene glycol ethers, their aldehyde and their acid metabolites were evaluated for their mutagenicity with the Ames test. The Salmonella typhimurium his- tester strains TA 97a, TA 98, TA 100 and TA 102 were used with and without rat S9
L Aasmoe et al.
Xenobiotica; the fate of foreign compounds in biological systems, 27(12), 1237-1244 (1998-02-14)
1. The elimination of 2-methoxyethanol (2-ME) and its toxic metabolite methoxyacetic acid (MAA) was studied in the male and female rat. We also studied the elimination of ethoxyacetic acid (EAA), the toxic metabolite formed by 2-ethoxyethanol (2-EE). 2. The rate
Esther de Jong et al.
Toxicological sciences : an official journal of the Society of Toxicology, 110(1), 117-124 (2009-04-30)
The embryonic stem cell test (EST) has been proposed as an in vitro assay that might reduce animal experimentation in regulatory developmental toxicology. So far, evaluation of the EST was not performed using compounds within distinct chemical classes. Evaluation within
T S Shih et al.
Occupational and environmental medicine, 56(7), 460-467 (1999-09-03)
To simplify the current preparation of samples, and to improve the specificity and reliability of the conventional analytical methods to measure urinary alkoxyacetic acids. Samples containing alkoxyacetic acids including methoxy, ethoxy, and butoxyacetic acids (MAA, EAA, and BAA) were acidified

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service