Skip to Content
MilliporeSigma
All Photos(1)

Documents

130400

Sigma-Aldrich

1,1,1-Trichlorotrifluoroethane

99%

Synonym(s):

CFC-113a

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl3CCF3
CAS Number:
Molecular Weight:
187.38
Beilstein:
1699455
EC Number:
MDL number:
UNSPSC Code:
12352103

vapor pressure

5.8 psi ( 20 °C)

Assay

99%

refractive index

n20/D 1.3599 (lit.)

bp

46 °C (lit.)

mp

13-14 °C (lit.)

density

1.579 g/mL at 25 °C (lit.)

SMILES string

FC(F)(F)C(Cl)(Cl)Cl

Application

Please view www.aldrich.com/epaods regarding the EPA′s request for application information of Ozone Depleting Substances

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Ozone 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service