Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

130087

Sigma-Aldrich

Myristic acid

95%

Synonym(s):

1-Tridecanecarboxylic acid, C14:0, NSC 5028, Tetradecanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)12COOH
CAS Number:
Molecular Weight:
228.37
Beilstein:
508624
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

Assay

95%

bp

250 °C/100 mmHg (lit.)

mp

52-54 °C (lit.)

functional group

carboxylic acid

lipid type

saturated FAs

shipped in

ambient

storage temp.

room temp

SMILES string

CCCCCCCCCCCCCC(O)=O

InChI

1S/C14H28O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14(15)16/h2-13H2,1H3,(H,15,16)

InChI key

TUNFSRHWOTWDNC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alicja Kuban-Jankowska et al.
PloS one, 7(12), e52495-e52495 (2013-01-10)
Hydrogen peroxide induces oxidation and consequently inactivation of many protein tyrosine phosphatases. It was found that hydrogen peroxide, in the presence of carboxylic acids, was efficiently activated to form even more potent oxidant - peroxy acid. We have found that
Geok-Lin Chua et al.
PloS one, 7(7), e41924-e41924 (2012-07-31)
Integrins are signal transducer proteins involved in a number of vital physiological processes including cell adhesion, proliferation and migration. Integrin molecules are hetero-dimers composed of two distinct subunits, α and β. In humans, 18 α and 8 β subunits are
Dalit Shental-Bechor et al.
Proceedings of the National Academy of Sciences of the United States of America, 109(44), 17839-17844 (2012-08-01)
We present an integrated experimental and computational study of the molecular mechanisms by which myristoylation affects protein folding and function, which has been little characterized to date. Myristoylation, the covalent linkage of a hydrophobic C14 fatty acyl chain to the
Anja Meier et al.
Hepatology (Baltimore, Md.), 58(1), 31-42 (2012-12-06)
Chronic infection with the human hepatitis B virus (HBV) is a global health problem and a main cause of progressive liver diseases. HBV exhibits a narrow host range, replicating primarily in hepatocytes. Both host and hepatocyte specificity presumably involve specific
S Curry et al.
Biochimica et biophysica acta, 1441(2-3), 131-140 (1999-11-26)
Human serum albumin possesses multiple fatty acid binding sites of varying affinities, but the precise locations of these sites have remained elusive. The determination of the crystal structure of human serum albumin complexed with myristic acid recently revealed the positions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service