Skip to Content
MilliporeSigma
All Photos(1)

Documents

125075

Sigma-Aldrich

2-Chloro-4-methylaniline

98%

Synonym(s):

2-Chloro-p-toluidine, 4-Amino-3-chlorotoluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(CH3)NH2
CAS Number:
Molecular Weight:
141.60
Beilstein:
774514
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.575 (lit.)

bp

223-225 °C (lit.)

mp

7 °C (lit.)

solubility

methanol: soluble

density

1.151 g/mL at 25 °C (lit.)

SMILES string

Cc1ccc(N)c(Cl)c1

InChI

1S/C7H8ClN/c1-5-2-3-7(9)6(8)4-5/h2-4H,9H2,1H3

InChI key

XGYLSRFSXKAYCR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Chloro-4-methylaniline was used in determination of aniline derivatives in environmental water samples using on-line combination of supported liquid membrane extraction and liquid chromatography. It was also used in determination of amines in environmental water samples by gas chromatography–mass spectrometry.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

210.2 °F - closed cup

Flash Point(C)

99 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fully automated on-line supported liquid membrane-liquid chromatographic determination of aniline derivates in environmental waters.
Norberg J, et al.
Chromatographia, 46(9-10), 483-488 (1997)
Miguel del Nogal Sánchez et al.
Analytical and bioanalytical chemistry, 404(6-7), 2007-2015 (2012-08-14)
We report a sensitive method for the determination of 15 aromatic amines in environmental water samples. They have been included in the list of priority pollutants in surface water by the European Union. The method is based on analyte enrichment

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service