Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

122580

Sigma-Aldrich

Stearonitrile

technical grade, ≥90%

Synonym(s):

Heptadecyl cyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)16CN
CAS Number:
Molecular Weight:
265.48
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

≥90%

form

solid

bp

274 °C/100 mmHg (lit.)

mp

38-40 °C (lit.)

density

0.818 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCCC#N

InChI

1S/C18H35N/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19/h2-17H2,1H3

InChI key

RHSBIGNQEIPSCT-UHFFFAOYSA-N

General description

Stearonitrile undergoes W-6 Raney nickel catalyzed hydrogenation reaction. It forms inclusion complexes with urea.

Application

Stearonitrile was used to study the behaviour of dipalmitoylphosphatidylcholine mixed with stearonitrile at the air–water interface by surface pressure–area measurements and by direct visualisation of monolayers by Brewster angle microscopy. It was used to study the phase behaviour of stearic acid mixed with stearonitrile, in bulk and in a monolayer at the air–water interface.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rute I S Romão et al.
Chemistry and physics of lipids, 131(1), 27-39 (2004-06-24)
The behaviour of dipalmitoylphosphatidylcholine (DPPC), mixed with stearonitrile (SN), was investigated at the air-water interface by surface pressure-area (pi-A) measurements and by direct visualisation of monolayers by Brewster angle microscopy (BAM). The pi-A-X diagram of system DPPC/SN was compared with
A C T Teixeira et al.
Chemistry and physics of lipids, 144(2), 160-171 (2006-10-10)
The solid-liquid phase behaviour of stearic acid (SA) and stearonitrile (SN) in binary mixtures was investigated by differential scanning calorimetry (DSC), and the formation of SA-SN mixed monolayers at the air-water interface was followed by surface pressure-area (pi-A) measurements and
A CP-MAS NMR study of n-alkanes included in urea: conformational dependences of the substituent effects for n-substituted alkanes.
Okazaki M and McDowell CA.
Journal of Molecular Structure, 118(1), 149-156 (1984)
The preparation of Raney nickel catalysts and their use under conditions comparable with those for platinum and palladium catalysts.
Adkins H and Billica HR.
Journal of the American Chemical Society, 70(2), 695-698 (1948)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service