Skip to Content
MilliporeSigma
All Photos(1)

Documents

118559

Sigma-Aldrich

1,5-Hexadien-3-ol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHCH2CH(OH)CH=CH2
CAS Number:
Molecular Weight:
98.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

refractive index

n20/D 1.448 (lit.)

density

0.878 g/mL at 25 °C (lit.)

SMILES string

OC(CC=C)C=C

InChI

1S/C6H10O/c1-3-5-6(7)4-2/h3-4,6-7H,1-2,5H2

InChI key

SZYLTIUVWARXOO-UHFFFAOYSA-N

General description

1,5-Hexadien-3-ol is the starting reagent for the enantioselective synthesis of (+)-rogioloxepane A. It undergoes Sharpless kinetic resolution and ring-closing metathesis to yield nine membered oxocene.

Application

1,5-Hexadien-3-ol was used in the synthesis of (+)-obtusenyne.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

84.2 °F - closed cup

Flash Point(C)

29 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael T Crimmins et al.
Journal of the American Chemical Society, 125(25), 7592-7595 (2003-06-19)
A total synthesis of the laurencia metabolite (+)-obtusenyne has been completed. The key steps include a Sharpless kinetic resolution and an asymmetric glycolate alkylation to establish the stereogenic centers adjacent to the ether linkage and a ring-closing metathesis reaction to
Induction of rejection of successful allografts of rat islets by donor peritoneal exudate cells.
P E Lacy et al.
Transplantation, 28(5), 415-420 (1979-11-01)
Yan-Lun Ju et al.
Plant physiology and biochemistry : PPB, 130, 501-510 (2018-08-11)
Grapes are one of the most important fruits because of their economic and nutritional benefits, and grapevines are widely cultivated in arid and semi-arid areas. Therefore, it is critical to study the mechanism by which grapevines respond to water stress.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service