Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

116556

Sigma-Aldrich

Harmol hydrochloride dihydrate

98%

Synonym(s):

1-Methyl-9H-pyrido(3,4-b)indol-7-ol, 1-Methyl-9H-pyrido[3,4-b]indol-7-ol hydrochloride dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H10N2O · HCl · 2H2O
CAS Number:
Molecular Weight:
270.71
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

SMILES string

Cl[H].[H]O[H].[H]O[H].Cc1nccc2c3ccc(O)cc3[nH]c12

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marcos Tascon et al.
Analytical and bioanalytical chemistry, 407(13), 3637-3645 (2015-03-10)
The use of algae as a foodstuff is rapidly expanding worldwide from the East Asian countries, where they are also used for medical care. Harmala alkaloids (HAlk) are a family of bioactive compounds found in the extracts of some plants

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service