Skip to Content
MilliporeSigma
All Photos(1)

Documents

110450

Sigma-Aldrich

1-Cyclopentylethanol

97%

Synonym(s):

α-Methylcyclopentanemethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(C5H9)OH
CAS Number:
Molecular Weight:
114.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

refractive index

n20/D 1.458 (lit.)

bp

167 °C (lit.)

density

0.919 g/mL at 25 °C (lit.)

SMILES string

CC(O)C1CCCC1

InChI

1S/C7H14O/c1-6(8)7-4-2-3-5-7/h6-8H,2-5H2,1H3

InChI key

WPCMSUSLCWXTKB-UHFFFAOYSA-N

General description

1-Cyclopentylethanol is synthesized from oxazaborolidines and methyllithium.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

144.0 °F - closed cup

Flash Point(C)

62.2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diane M Hinkens et al.
The Journal of organic chemistry, 74(11), 4143-4148 (2009-05-13)
Dicyclohexyl-6-iodohexenylborane efficiently undergoes radical cyclization at room temperature using tri-n-butyltin hydride as a hydrogen donor and without the aide of a radical initiator. Efforts to develop environmentally friendly reagents using hypophosphites as substitutes for tin hydrides in this reaction provided

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service