Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

110159

Sigma-Aldrich

Vinylcyclohexene dioxide

97%

Synonym(s):

1,2-Epoxy-4-epoxyethylcyclohexane, 3-Epoxyethyl-7-oxabicyclo[4.1.0]heptane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12O2
CAS Number:
Molecular Weight:
140.18
Beilstein:
106071
EC Number:
MDL number:
UNSPSC Code:
12162002

Assay

97%

refractive index

n20/D 1.477 (lit.)

bp

230-232 °C (lit.)

density

1.094 g/mL at 25 °C (lit.)

SMILES string

C1CC2OC2CC1C3CO3

InChI

1S/C8H12O2/c1-2-6-7(10-6)3-5(1)8-4-9-8/h5-8H,1-4H2

InChI key

OECTYKWYRCHAKR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service