Skip to Content
MilliporeSigma
All Photos(1)

Documents

109002

Sigma-Aldrich

Sodium thioglycolate

97%

Synonym(s):

Mercaptoacetic acid sodium salt, Thioglycolic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSCH2COONa
CAS Number:
Molecular Weight:
114.10
Beilstein:
4569109
EC Number:
MDL number:
UNSPSC Code:
12352103

Assay

97%

mp

>300 °C (lit.)

SMILES string

[Na+].[O-]C(=O)CS

InChI

1S/C2H4O2S.Na/c3-2(4)1-5;/h5H,1H2,(H,3,4);/q;+1/p-1

InChI key

GNBVPFITFYNRCN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

For preparation of bacteriological media. Analytical reagent.

replaced by

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service