Skip to Content
MilliporeSigma
All Photos(1)

Documents

102938

Sigma-Aldrich

2-Acetyl-5-norbornene, mixture of endo and exo

technical grade

Synonym(s):

Methyl 5-norbornen-2-yl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H12O
CAS Number:
Molecular Weight:
136.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

refractive index

n20/D 1.484 (lit.)

bp

84-86 °C/18 mmHg (lit.)

density

1.005 g/mL at 25 °C (lit.)

SMILES string

CC(=O)C1C[C@H]2C[C@@H]1C=C2

InChI

1S/C9H12O/c1-6(10)9-5-7-2-3-8(9)4-7/h2-3,7-9H,4-5H2,1H3/t7-,8+,9?/m1/s1

InChI key

NIMLCWCLVJRPFY-WGTSGOJVSA-N

General description

2-Acetyl-5-norbornene, mixture of endo and exo is a colorless to light yellow liquid.

Application

2-Acetyl-5-norbornene was used in the synthesis of the 2-acetylnorbornyl isothiocyanates: exo-2-acetyl-exo-6-isothiocyanatonorbornane, endo-2-acetyl-exo-6-isothiocyanatonorbornane and exo-2-acetyl-exo-5-isothiocyanatonorbornane.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

143.6 °F - closed cup

Flash Point(C)

62 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Zhang et al.
Proceedings of the National Academy of Sciences of the United States of America, 91(8), 3147-3150 (1994-04-12)
Sulforaphane [1-isothiocyanato-4-(methyl-sulfinyl)butane] was recently isolated from one variety of broccoli as the major and very potent inducer of phase 2 detoxication enzymes in murine hepatoma cells in culture. Since phase 2 enzyme induction is often associated with reduced susceptibility of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service