Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AC7300013717

Advanced Chemical Intermediates Ltd.

5-Pyridin-2-yl-thiophene-2-sulfonyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
259.74
MDL number:

Iupac Name

5-(2-pyridinyl)-2-thiophenesulfonyl chloride

Inchi Code

1S/C9H6ClNO2S2/c10-15(12,13)9-5-4-8(14-9)7-3-1-2-6-11-7/h1-6H

InChI key

OGOMWPWAEIDEOU-UHFFFAOYSA-N

storage temp.

Room temperature

Purity

95%

Country of Origin

GB

Shipping Temperature

Room temperature

Physical Form

off white solid

Product Link

https://www.acints.com/products/ACI-01115

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

MSDS

https://www.acints.com/products/ACI-01115


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service