Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ADVH95EC9C4F

AChemBlock

3-methylazetidin-2-one

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€628.30
1 G
€1,205.80
2 G
€2,188.30

Vendor SKU: 12L-716-500MG

€628.30


Ships in 21 days - from Aldrich Partner.


Select a Size

Change View
500 MG
€628.30
1 G
€1,205.80
2 G
€2,188.30

About This Item

CAS Number:
Molecular Weight:
85.11
MDL number:

Vendor SKU: 12L-716-500MG

€628.30


Ships in 21 days - from Aldrich Partner.

Iupac Name

3-methyl-2-azetidinone

Inchi Code

1S/C4H7NO/c1-3-2-5-4(3)6/h3H,2H2,1H3,(H,5,6)

InChI key

XYAPRMQTKAOTJZ-UHFFFAOYSA-N

storage temp.

Store at 0-8 C

Purity

97%

Country of Origin

CN

Physical Form

light yellow liquid

Product Link

https://www.achemblock.com/s74937-3-methylazetidin-2-one.html

Synonym

3-methylazetidin-2-one

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://www.achemblock.com/s74937-3-methylazetidin-2-one.html


  • Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    Sorry, we don't have COAs for this product available online at this time.

    If you need assistance, please contact Customer Support.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service