Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ADVH303C3399

AChemBlock

3-bromo-4-chloro-2-hydroxybenzaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
235.46
MDL number:

Iupac Name

3-bromo-4-chloro-2-hydroxybenzaldehyde

Inchi Code

1S/C7H4BrClO2/c8-6-5(9)2-1-4(3-10)7(6)11/h1-3,11H

InChI key

IESXXBIVEXVFRF-UHFFFAOYSA-N

storage temp.

Store at 0-8 C

Purity

95%

Country of Origin

CN

Physical Form

brown powder

Product Link

https://www.achemblock.com/q69467-3-bromo-4-chloro-2-hydroxybenzaldehyde.html

Synonym

3-bromo-4-chloro-2-hydroxybenzaldehyde

Pictograms

Exclamation mark

Signal Word

warning

Hazard Statements

Precautionary statements

MSDS

https://www.achemblock.com/q69467-3-bromo-4-chloro-2-hydroxybenzaldehyde.html


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service