Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AC5372910047

Accela ChemBio Inc.

7-Methyl-1,3-dihydroimidazo[4,5-b]pyridin-2-one

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
149.15
MDL number:

Iupac Name

7-methyl-3H-imidazo[4,5-b]pyridin-2-ol

Inchi Code

1S/C7H7N3O/c1-4-2-3-8-6-5(4)9-7(11)10-6/h2-3H,1H3,(H2,8,9,10,11)

InChI key

OPDZDXBVPWMSKU-UHFFFAOYSA-N

storage temp.

RT

Purity

95%

Country of Origin

CN

Shipping Temperature

RT

Physical Form

Brown or reddish-brown solid

Product Link

http://www.accelachem.com/cn/search.html?keyword=SY007154&attname=GoodsCode

MSDS

http://pdf.accelachem.com/cn/msds/SY007154.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service