Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AABH9616B679

AA BLOCKS, INC.

Diacetato[(S)-(-)-2,2'-bis(diphenylphosphino)-5,5',6,6',7,7',8,8'-octahydro-1,1'-binaphthyl]ruthenium(II)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
1581.73
MDL number:

Iupac Name

bis(λ²-ruthenium(2+)) bis([2'-(diphenylphosphanyl)-5H,5'H,6H,6'H,7H,7'H,8H,8'H-[1,1'-binaphthalen]-2-yl]diphenylphosphane) diacetate

Inchi Code

1S/2C44H40P2.2C2H4O2.2Ru/c2*1-5-19-35(20-6-1)45(36-21-7-2-8-22-36)41-31-29-33-17-13-15-27-39(33)43(41)44-40-28-16-14-18-34(40)30-32-42(44)46(37-23-9-3-10-24-37)38-25-11-4-12-26-38;2*1-2(3)4;;/h2*1-12,19-26,29-32H,13-18,27-28H2;2*1H3,(H,3,4);;/q;;;;2*+2/p-2

InChI key

RFJUJGMACYPAMB-UHFFFAOYSA-L

storage temp.

2~8C

Country of Origin

CN

Product Link

https://www.aablocks.com/prod/142962-95-6

Synonym

Diacetato[(S)-(-)-2,2'-bis(diphenylphosphino)-5,5',6,6',7,7',8,8'-octahydro-1,1'-binaphthy]ruthenium(II)

MSDS

https://www.aablocks.com/downloadSds?cas=142962-95-6


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service