MilliporeSigma
Dual-Labeled Probes
  • Amounts: 1, 3, 5, and 10 OD
  • Purification: HPLC
  • Sequence Lengths: 15 - 40 bases
  • Modifications: Locked Nucleic Acid available
  • Dyes: Comprehensive offering of reporters and quenchers
  • Quality Control: 100% mass spectrometry
  • Format: Supplied dry in amber tubes
  • Learn more
Molecular Beacons
  • Amounts: 1, 3, 5, and 10 OD
  • Purification: HPLC
  • Sequence Lengths: 15 - 40 bases
  • Modifications: Locked Nucleic Acid available
  • Dyes: Comprehensive offering of reporters and quenchers
  • Quality Control: 100% mass spectrometry
  • Format: Supplied dry in amber tubes
  • Learn more
LightCycler® Probes
  • Amounts: 0.1, 0.25, 1.5, and 15 OD
  • Purification: HPLC
  • Sequence Lengths: 15 - 40 bases
  • Modifications: Locked Nucleic Acid available
  • Dyes: Probe 1 fluorescein (3′ end); Probe 2 phosphate (3′ end) and LC Red 610, 640, 670 or 705 (5′ end)
  • Quality Control: 100% mass spectrometry
  • Format: Supplied dry in amber tubes
  • Learn more
Scorpions® Probes
  • Amounts: 1, 5, and 5 OD
  • Purification: HPLC
  • Sequence Lengths: 30 - 60 bases
  • Modifications: Locked Nucleic Acid available
  • Dyes: Comprehensive offering of reporters and quenchers
  • Quality Control: 100% mass spectrometry
  • Format: Supplied dry in amber tubes
  • Learn more
Locked Nucleic Acid
  • Available for all Custom qPCR Probe Types
  • Increased thermal stability and hybridization specificity
  • Easier and more specific designs for problematic target sequences
  • Learn more
Black Hole Quencher™ (BHQ™)
  • Available for all Custom qPCR Probe types
  • No native fluorescence (emits heat instead of light), resulting in lower background fluorescence
  • Increased signal-to-noise ratio, providing higher sensitivity
  • Enables a wider choice of reporters for multiplexing
  • Learn more