MilliporeSigma
HomeWebinarsSPME in Food Analysis with New Coating
SPME in Food Analysis with New Coating
WEBINAR
Description / Abstract
The complexity and diversity food matrices and the need for clean extracts to be injected into analytical instrumentation may imply the use of tedious and time-consuming sample preparation strategies that often are a source of a significant amount of laboratory waste. The development of a new matrix-compatible solidphase microextraction (SPME) coating, namely PDMS/DVB/PDMS, bearing enhanced antifouling properties, enabled the analysis of complex food matrices by direct immersion SPME and helped to overcome issues related to extensive sample pre-treatment and instrumental contamination. This webinar will focus on recent advances made toward the analysis of contaminants in complex food samples by using the new matrix-compatible SPME coating and will describe the optimization of important parameters to be considered when performing DI-SPME.
Speaker
Emanuela Gionfriddo
Emanuela Gionfriddo
University of Waterloo
Research Associate
Emanuela Gionfriddo is currently a Research Associate in Prof. Pawliszyn’s research group and manager of the Gas-Chromatography section of the Industrially Focused Analytical Research Laboratory (InFAReL) at University of Waterloo (ON, Canada). She received her B.Sc. (2008) and M.Sc. (2010) in Chemistry and her Ph.D. in Analytical Chemistry (2013) at University of Calabria (Italy). Since 2014, she has been working in Prof. Pawliszyn’s research group: her research focuses on the development and testing of new matrix compatible SPME coatings, compatible with both gas- and liquid- chromatography applications and suitable for direct immersion in complex matrices such as biological fluids and foodstuffs: potential application of these coatings involves the coupling of complementary chromatography platforms for both targeted multiresidue analysis and metabolomics studies. She is also active in the development of new SPME methods for determination of contaminants at trace level in various food, environmental matrices and pharmaceutical supplements by GC-MS, GC x GC-MS and direct coupling with MS.
Webinar Information
Food and beverage testing and manufacturing
  • Chemical analysis
  • Duration:1h 5min
  • Language:English