Matrix-rich samples often require extensive sample preparation when analyzed with HPLC and UHPLC. This is a substantial time and cost factor. A robust column material enabling a simplified sample preparation will save time and improve the lab productivity.
The performance of HPLC columns has improved dramatically in recent years, particularly in terms of separation power as measured by the number of theoretical plates per meter. The improvement in performance has been achieved primarily by a reduction in particle size.
Not all samples have benefited equally from this development: Clean samples with a relatively simple matrix are ideally suited for this new generation of columns. But for samples with a more difficult and complex matrix, the benefits are often accompanied by significant limitations. The explanation is quite simple; the smaller the particle size and the higher the sample amount injected, the more easily a column plugs or blocks. If the sample contains a matrix with colloidal and micro-particulate matter in suspension, then it becomes necessary to perform a rigorous sample “clean-up” before injection. This demands additional time for sample preparation steps, and of course involves additional cost.
The potential for cost and time savings by increasing lab productivity with faster separations and column materials providing very high matrix tolerance will be discussed.
By attending this presentation you will...
- Understand the challenges for matrix-rich sample analysis using HPLC and/or UHPLC and how to overcome them
- Learn how to leverage lab efficiency by simplifying the HPLC workflow using monolithic silica columns.
Speakers
![Petra Lewits](/deepweb/assets/sigmaaldrich/marketing/global/images/webinars/petra-lewits/petra-lewits.jpg)
Petra Lewits
MilliporeSigma
Global Product Manager for Analytical Chromatography, HPLC columns
Petra has worked with chromatography for 40 years. She started her career in separation science in 1983 using several chromatographic methods, including HPLC, TLC, GC and solid-phase extraction (SPE), for food control in a governmental institution lab. In 1986 she joined the pharmaceutical R&D and QC department (Merck KGaA, Darmstadt, Germany) where she developed HPLC methods for R&D and QC of pharmaceutical APIs and formulations as well as for diet convenience foods. From 1991 on she supported customers in different positions with technical and application-related inquires in Europe as a sales specialist for HPLC instruments, also providing technical support for chromatography and complaint management. Since 2002 she has been a Global Product Manager for Analytical Chromatography (HPLC). In 2014 she began focusing on thin-layer chromatography (TLC). Since April 2018 Petra has had global responsibility for analytical HPLC columns.
Analytical chemistry
- Low pressure liquid chromatography
Duration:1h
Language:English
Session 1:presented December 2, 2021