MilliporeSigma
HomeProfessor Product PortalMaruoka Group – Professor Product Portal
Maruoka Group – Professor Product Portal
Professor Keiji Maruoka
Professor Keiji Maruoka
Professor Keiji Maruoka and co-workers have developed a series of designer chiral phase-transfer catalysts that are useful for various asymmetric transformations including asymmetric alkylation, aldol reaction, Mannich reaction, conjugate addition, epoxidation, and Strecker reaction. In particular, “Maruoka Catalysts®” are useful for the large-scale production of various artificial amino acids through the asymmetric alkylation of glycine, alanine, and other peptide derivatives. Prof. Maruoka is also known for his work with the design of high-performance organocatalysts for efficient asymmetric catalysis. Some of these have been commercialized, including 3,3’-diaryl-substituted binaphthyldicarboxylic acids as chiral Brønsted acid catalysts.
Maruoka Group Website
Recent papers from the Maruoka Group
1.
Shirakawa S, Maruoka K. 2013. Recent Developments in Asymmetric Phase-Transfer Reactions. Angew. Chem. Int. Ed.. 52(16):4312-4348. https://doi.org/10.1002/anie.201206835
Loading
DISCIPLINES
  • Organic Synthetic Chemistry
  • Catalysis
Professor Product Portal Index