MilliporeSigma
HomeProfessor Product PortalCarlson Group – Professor Product Portal
Carlson Group – Professor Product Portal
Erin Carlson
Professor Erin Carlson
Most bacteria are surrounded by a cell wall composed of a complex polymeric structure called peptidoglycan that is essential for cell survival. The biosynthetic pathway for production of peptidoglycan and the proteins required for its assembly have been the targets for many antibacterial agents. For example, penicillin-binding proteins (PBPs), which polymerize and crosslink strands of peptidoglycan also have affinity for the beta-lactam antibiotic penicillin. PBPs are classified into three groups based upon their molecular weight, conserved amino acid motifs and function. These proteins all contain a serine in their peptidase domain that is required for catalysis. Penicillin hinders PBP function by forming a stable acyl-enzyme intermediate with this residue, which in turn inhibits crosslinking of peptidoglycan.
Despite the effectiveness of beta-lactam antibiotics, bacterial resistance has arisen very rapidly. A more detailed understanding of the mechanism of peptidoglycan synthesis may be the key for design of new and more effective antibiotics. We have developed fluorescently-labeled small molecule probes to visualize the activity of PBPs directly in live cells. Small molecule-conjugated fluorophores can provide superior temporal resolution than more traditional strategies, such as protein fusion, and their activity can be modulated by dose. These compounds are also generally easy to use and enable visualization in a broad range of organisms to explore the construction and remodeling of peptidoglycan.
Carlson Group Website
Recent papers from the Carlson Group
1.
Kocaoglu O, Calvo RA, Sham L, Cozy LM, Lanning BR, Francis S, Winkler ME, Kearns DB, Carlson EE. 2012. Selective Penicillin-Binding Protein Imaging Probes Reveal Substructure in Bacterial Cell Division. ACS Chem. Biol.. 7(10):1746-1753. https://doi.org/10.1021/cb300329r
2.
Sharifzadeh S, Boersma MJ, Kocaoglu O, Shokri A, Brown CL, Shirley JD, Winkler ME, Carlson EE. 2017. Novel Electrophilic Scaffold for Imaging of Essential Penicillin-Binding Proteins inStreptococcus pneumoniae. ACS Chem. Biol.. 12(11):2849-2857. https://doi.org/10.1021/acschembio.7b00614
3.
Watt VM, Yip CC. 1989. HeLa cells contain the atrial natriuretic peptide receptor with guanylate cyclase activity. Biochemical and Biophysical Research Communications. 164(2):671-677. https://doi.org/10.1016/0006-291x(89)91512-x
DISCIPLINES
  • Organic Synthetic Chemistry
  • Catalysis
Professor Product Portal Index