MilliporeSigma
The Wait Is Over
Milliflex® Rapid System speeds up your microbial QC
SPEED UP YOUR MICROBIAL QC TESTING WITH A PROVEN RAPID MICROBIOLOGY METHOD
Milliflex® Rapid System 2.0 is an automated system for rapid and accurate detection of contaminant microorganisms. Suitable for rapid sterility, bioburden, in-process, and product release testing, the Milliflex® Rapid System 2.0 is equipped with a range of new features but is still based on the same proven ATP-bioluminescence technology and offers the same performance. Ready to get your microbial results faster?
Watch the workflow video to learn how to speed up your time-to-result with the Milliflex® Rapid system 2.0.
Key benefits:
  • Consistently fast and reproducible results
  • Up to four times faster results than traditional membrane filtration or pour plate methods
  • CFU counts correlate with those of traditional methods
  • Data integrity thanks to 21 CFR Part 11 compliant software
  • Easy handling and validation
Extended service offering including validation support
 For more information contact us, view our related products, or explore our related resources.
Attend our webinar to learn from our experts how you can benefit from using the new rapid microbial detection platform.
A Few Simple Steps to Speed Up Your Microbial Testing Workflow
Step 1: Membrane-Based Sample Preparation
Milliflex® Membrane-Based Sample Preparation
    Step 2: Optimized Reagent Spraying
    Milliflex® Optimized Reagent Spraying
      Step 3: Automated Microorganisms Enumeration
      Milliflex® Automated Microorganisms Enumeration
        Related Products
        Loading
        Related Resources
        Request a Demo
        Fields indicated by an * are required.