MilliporeSigma
Cell Monitoring, Evolved
Millicell® DCI Digital Cell Imager
Millicell® DCI Digital Cell Imager
Watch this video to see how UNSW leverages the Millicell® DCI in their research.
Assess your cell cultures with ease. The Millicell® DCI Digital Cell Imager enables more efficient execution of the repetitive daily techniques associated with cell passaging.
Take Confluency To A New Level
  • Quickly and objectively measure common cell culture parameters including confluency, cell count, and morphology.
  • Monitor an extensive range of adherent cell cultures including 3D cultures and tricky or hard-to-grow cells.
  • Save time and conserve precious culture sample with in-vessel measurement.
  • Track and record cell culture data using streamlined web-based data management tools.
  • Analyze cell growth trends with instant access to historical data for more consistent cell cultures.
Related Products
Loading
Increase lab productivity
Expand your lab’s cell culturing capacity with the Millicell® DCI.
  • Faster analysis
  • Reduced user bias
  • Accurate results
  • More consistent cell cultures
  • Confluency, cell count, and morphology
  • Hemocytometer or in-vessel measurement
  • Two-click image capture
  • Convenient, web-based cloud service for data analysis, storage, and archiving
Accelerate Cell Culture Analysis
Accelerate cell culture analysis
With an intuitive interface that automates analysis, the Millicell® DCI streamlines cell culture measurements. From convenient slide bars that allow you to adjust measurements based on cell size and masking area to automatic grid finders for hemocytometer measurements, the Millicell® DCI was designed for experts and novices, alike.
Expand access and capability
Expand access and capability
Our web-based application allows for convenient downstream analysis in addition to sorting and recording data and images. This subscription service can be used to save costs and support data redundancy for backup and protection. Meet your regulatory requirements with secure, cloud-based data storage and archiving.
Latest Software
ALREADY HAVE AN INSTRUMENT?
Download our latest software to take full advantage of new features and updates.
Related Technical Resources