Long Oligos
Our experience with gene construction and microarray development provides us with insight into the potential difficulties of long-oligo synthesis. The techniques we have developed to purify long oligos are unmatched by other suppliers.
Long Oligo Specifications | |
---|---|
Bases | DNA (A,C,G,T) and degenerate bases |
Backbone | Phosphodiester and phosphorothioates |
Length | 121-180 bases |
Yield | Cartridge: 5 OD, PAGE: 0.5 OD |
Purification | Cartridge or PAGE |
Modifications / Modified Bases | 5' end: Phos, Biotin, Amine C6, Amine C12, Inosine, deoxyUracil 3' end: Spacer C3 Internal: Inosine and deoxyUracil |
Format | Dry in 2 mL screw cap tubes |
Documentation | All oligos are delivered with Quality Assurance document including yields, Tm (melting temperature), MW, and µg/OD |
Quality Control | Every oligo is quality checked by mass spectrometry or PAGE Gel Analysis and OD by UV Spectroscopy |
Shipping Schedule | Cartridge and PAGE: 10 business days |
Products
Sorry, an unexpected error has occurred
Network error: Failed to fetch