GC Analysis of Patchouli Essential Oil on SLB®-5ms
![GC Analysis of Patchouli Essential Oil on SLB®-5ms suitable for GC](/deepweb/assets/sigmaaldrich/product/images/130/683/e2b37da8-237a-4bed-a15e-1fa9c332c071/800/e2b37da8-237a-4bed-a15e-1fa9c332c071.jpg)
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
SLB-5ms, 30 m x 0.25 mm I.D., 0.25 μm (28471-U)
oven
50 °C (0.5 min), then 5 °C/min to 300 °C (10 min)
inj. temp.
280 °C
detector
FID, 310 °C
carrier gas
helium, 25cm/ sec set at 110°C
injection
0.1 μL 100:1 split
liner
4 mm I.D. cup injector
sample
Patchouli Oil, neat (W283800)
Description
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany